Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTNA Monoclonal Antibody (OTI2F9), TrueMAB™, OriGene
SDP

Catalog No. 502541676
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1676 30 μL
50-167-3330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1676 Supplier OriGene Supplier No. TA502258S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DTNA Monoclonal antibody specifically detects DTNA in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen DTNA
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2F9
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TASP1
Gene Accession No. Q9H6P5
Gene Alias 2210407P21Rik; a-DB-1; adbn; Alpha-dystrobrevin; D18S892E; DRP3; Dtn; Dtna; DTN-A; Dtna-ps1; dystrobrevin alpha; dystrobrevin alpha, pseudogene 1; dystrobrevin, alpha; dystrophin-related protein 3; Gm19389; LVNC1; RGD1561985
Host Species Mouse
Immunogen Full length human recombinant protein of human TASP1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55617
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.