Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTNA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588329 Supplier Invitrogen™ Supplier No. PA588329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVECSYCHSE SMMGFRYRCQ QCHNYQLCQD CFWRGHAGGS HSNQHQMKEY TSWKSPAKKL TNALSKSLSC ASSREPLHPM FPDQPEKPLN LAHIVDTWPP RPVTSMNDTL FSHSVPSSGS PFITRSSPPK DSEVEQNKLL ARAAPAFLKG KGIQYSLNVA DRLADEHVLI GLYVNMLRNN PSCMLESSNR LDEEHRLIAR YAARLAAESS SSQPPQQRSA PDISFTIDAN KQQRQLIAEL E; Positive Samples: U-87MG, Mouse brain; Cellular Location: Cell junction, Cell membrane, Cytoplasm, synapse.

The protein encoded by this gene belongs to the dystrobrevin subfamily of the dystrophin family. This protein is a component of the dystrophin-associated protein complex (DPC), which consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and alpha- and beta-dystrobrevin. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Mutations in this gene are associated with left ventricular noncompaction with congenital heart defects. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTNA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DTNA
Gene Accession No. Q9D2N4, Q9Y4J8
Gene Alias 2210407P21Rik; a-DB-1; adbn; Alpha-dystrobrevin; D18S892E; DRP3; Dtn; Dtna; DTN-A; Dtna-ps1; dystrobrevin alpha; dystrobrevin alpha, pseudogene 1; dystrobrevin, alpha; dystrophin-related protein 3; Gm19389; LVNC1; RGD1561985
Gene Symbols DTNA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-480 of human DTNA (NP_0013812).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13527, 1837
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.