Learn More
Invitrogen™ DTNBP1 Polyclonal Antibody

Description
Immunogen sequence: GLKTLSDKSR EAKVKSKPRT VPFLPKYSAG LELLSRYEDT WAALHRRAKD CASAGELVDS EVVMLSAHWE KK Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 90%.
Specifications
Specifications
| Antigen | DTNBP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DTNBP1 |
| Gene Accession No. | Q96EV8 |
| Gene Alias | 5430437B18Rik; AW048963; biogenesis of lysosomal organelles complex-1, subunit 8; biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; Bloc1s8; DBND; distrobrevin binding protein 1; DKFZp564K192; DTBP1; Dtnbp1; Dysbindin; dysbindin-1; dystrobrevin binding protein 1; dystrobrevin-binding protein 1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; hermansky-Pudlak syndrome 7 protein homolog; HPS7; HPS7 protein; HPS7 protein homolog; Hps7-like protein; MGC20210; My031; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1; sandy; sdy |
| Gene Symbols | DTNBP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.