Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTNBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102450 Supplier Invitrogen™ Supplier No. PA5102450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DTNBP1.

Dtnbp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTNBP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DTNBP1
Gene Accession No. Q5M834, Q91WZ8, Q96EV8
Gene Alias 5430437B18Rik; AW048963; biogenesis of lysosomal organelles complex-1, subunit 8; biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; Bloc1s8; DBND; distrobrevin binding protein 1; DKFZp564K192; DTBP1; Dtnbp1; Dysbindin; dysbindin-1; dystrobrevin binding protein 1; dystrobrevin-binding protein 1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; hermansky-Pudlak syndrome 7 protein homolog; HPS7; HPS7 protein; HPS7 protein homolog; Hps7-like protein; MGC20210; My031; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1; sandy; sdy
Gene Symbols DTNBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DTNBP1(Accession Q96EV8), corresponding to amino acid residues Q284-T334.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641528, 84062, 94245
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.