Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Grade DTT, Promega

Catalog No. PRP1171 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-P1171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-P1171 Supplier Promega Corporation Supplier No. P1171
Add to Cart
Add to Cart

An antioxidant used to stabilize enzymes and other proteins containing sulfhydryl groups.

  • Quality tested and certified free of DNase, RNase and protease activity
  • Available as a liquid (100μL) or as a powder (5g or 25g)
  • Supplied at a concentration of 100mM

Chemical Identifiers

CAS 12-3-3483
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-dtt, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, 1,4-dithiothreithol
PubChem CID 446094
ChEBI CHEBI:42170

Specifications

Color White
Quantity 100 μL
Formula Weight 154.25
Physical Form Crystals, Powder or Liquid
Chemical Name or Material DTT
Recommended Storage −30 to −10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.