Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTWD1 Monoclonal Antibody (OTI2C2)
GREENER_CHOICE

Catalog No. PIMA527492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527492 Supplier Invitrogen™ Supplier No. MA527492
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DTWD1 Monoclonal Antibody for Western Blot

Catalyzes the formation of 3-(3-amino-3-carboxypropyl)uridine (acp3U) at position 20 in the D-loop of several cytoplasmic tRNAs (acp3U(20)). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTWD1
Applications Western Blot
Classification Monoclonal
Clone OTI2C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DTWD1
Gene Accession No. Q8N5C7
Gene Alias DTW domain containing 1; DTW domain-containing protein 1; DTWD1; MDS009; x 009 protein
Gene Symbols DTWD1
Host Species Mouse
Immunogen Full length human recombinant protein of DTWD1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56986
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.