Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTWD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565902
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA565902 100 μL
1 options

Catalog No. PIPA565902

Supplier: Invitrogen™ PA565902

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EARTLQEPVAR PSGASSSQTP NDKERREGGA VPAAAALGAE ADDDSADGLW ELPVEPAERR PECTRCSRPQ KVCLCPFLPA Highest antigen sequence identity to the following orthologs - mouse 68%, rat 68%.

DTWD2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTWD2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DTWD2
Gene Accession No. Q8NBA8
Gene Alias 1190002H09Rik; 8030470C17Rik; AI414673; BB115449; DTW domain containing 2; DTW domain-containing protein 2; Dtwd2
Gene Symbols DTWD2
Host Species Rabbit
Immunogen Recombinant Human DTWD2. Recombinant protein control fragment (Product #RP-105003).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285605
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.