Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTX1 Polyclonal Antibody

Catalog No. PIPA522227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522227 Supplier Invitrogen™ Supplier No. PA522227

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (98%), Xenopus laevis (86%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Studies in Drosophila have identified this gene as encoding a positive regulator of the Notch-signaling pathway. The human gene encodes a protein of unknown function; however, it may play a role in basic helix-loop-helix transcription factor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Dtx1
Gene Accession No. Q86Y01
Gene Alias deltex 1; deltex 1 homolog (Drosophila); deltex 1, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 1; deltex homolog 1; deltex1; DTX1; E3 ubiquitin-protein ligase DTX1; fractionated x-irradiation induced transcript 1; Fxit1; FXI-T1; hDTX1; hDx-1; mDTX1; mKIAA4160; protein deltex-1; RING-type E3 ubiquitin transferase DTX1
Gene Symbols Dtx1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Deltex1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1840
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.