Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTX2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152144 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152144 Supplier Novus Biologicals Supplier No. NBP152144
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

DTX2 Polyclonal specifically detects DTX2 in Human, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen DTX2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_065943.2
Gene Alias deltex (Drosophila) homolog 2, deltex homolog 2 (Drosophila), Deltex2, hDTX2, MGC71098, protein deltex-2, RING finger protein 58, RNF58KIAA1528deltex2, zinc ion binding protein
Gene Symbols DTX2
Host Species Goat
Immunogen Peptide with sequence C-EKLSTASGYSDVTD corresponding to internal region according to NP_065943.2, NP_001096066.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 113878
Test Specificity This antibody is expected to recognize both reported isoforms (NP_065943.2; NP_001096066.1). Reported variants NP_001096065.1, NP_065943.2, NP_001096064.1 represent identical protein.
Target Species Human, Rat, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.