Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DTX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560164
Description
Immunogen sequence: YPVTTIIAPP GHTGVACSCH QCLSGSRTGP VSGRYRHSMT NLPAYPVPQH PPHRTASVFG THQAFAPYNK PSL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 75%.
DTX2 functions as an E3 ubiquitin ligase. Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Mediates the antineural activity of Notch, possibly by inhibiting the transcriptional activation mediated by MATCH1. Functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.
Specifications
DTX2 | |
Polyclonal | |
Unconjugated | |
DTX2 | |
2610524D08Rik; AA408415; AU022494; deltex 2 homolog; deltex 2 homolog (Drosophila); deltex 2, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 2; deltex homolog 2; deltex2; Dtx2; hDTX2; KIAA1528; mDTX2; probable E3 ubiquitin-protein ligase DTX2; Protein deltex-2; ring finger protein 58; RING-type E3 ubiquitin transferase DTX2; RNF58; zinc ion binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113878 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UW9 | |
DTX2 | |
Recombinant protein corresponding to Human DTX2. Recombinant protein control fragment (Product #RP-92627). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction