Learn More
Invitrogen™ DTX3 Monoclonal Antibody (4F10)

Description
Peptide Sequence: MCGRFYGQLV GNQPQNGRML VSKDATLLLP SYEKYGTIVI QYVFPPGVQG AEHPNPGVRY PGTTRVAYLP DCPEGNKVLT LFRKAFDQRL TFTIGTSMTT.
Specifications
Specifications
| Antigen | DTX3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4F10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | DTX3 |
| Gene Accession No. | Q8N9I9 |
| Gene Alias | deltex 3 homolog (Drosophila); deltex 3, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 3; deltex homolog 3; deltex3; DTX3; FLJ34766; mDTX3; mg310; MGC138863; MGC138864; probable E3 ubiquitin-protein ligase DTX3; Protein deltex-3; RING finger protein 154; RING-type E3 ubiquitin transferase DTX3; RNF154 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.