Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519212
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519212 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519212 Supplier Invitrogen™ Supplier No. PA519212
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTX3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DTX3
Gene Accession No. Q80V91, Q8N9I9
Gene Alias deltex 3 homolog (Drosophila); deltex 3, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 3; deltex homolog 3; deltex3; DTX3; FLJ34766; mDTX3; mg310; MGC138863; MGC138864; probable E3 ubiquitin-protein ligase DTX3; Protein deltex-3; RING finger protein 154; RING-type E3 ubiquitin transferase DTX3; RNF154
Gene Symbols DTX3
Host Species Goat
Immunogen Synthetic peptide sequence (KNKVTVSKPVWD) corresponding to the internal amino acids of DTX3 (aa 16-27).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196403, 80904
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.