Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563762 Supplier Invitrogen™ Supplier No. PA563762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPPRLREEAE EQESTCPICL GEIQNAKTLE KCRHSFCEGC ITRALQVKKA CPMC Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Probably acts both as a positive and negative regulator of Notch, depending on the developmental and cell context. Functions as an ubiquitin ligase protein in vitro, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTX3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DTX3
Gene Accession No. Q8N9I9
Gene Alias deltex 3 homolog (Drosophila); deltex 3, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 3; deltex homolog 3; deltex3; DTX3; FLJ34766; mDTX3; mg310; MGC138863; MGC138864; probable E3 ubiquitin-protein ligase DTX3; Protein deltex-3; RING finger protein 154; RING-type E3 ubiquitin transferase DTX3; RNF154
Gene Symbols DTX3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DTX3. Recombinant protein control fragment (Product #RP-101968).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196403
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.