Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DTX3L Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002873AF488
Description
DTX3L Polyclonal antibody specifically detects DTX3L in Human samples. It is validated for Western BlotSpecifications
DTX3L | |
Polyclonal | |
Western Blot | |
BBAPEC 6.3.2.-, B-lymphoma- and BAL-associated protein, deltex 3-like (Drosophila), E3 ubiquitin-protein ligase DTX3L, Protein deltex-3-like, rhysin 2, rhysin2, Rhysin-2 | |
The immunogen recognized by this antibody maps to a region between residue 700 and the C-terminus (residue 740) of human Deltex 3-Like Protein (B-Lymphoma- and BAL-Associated Protein) using the numbering given in entry NP_612144.1 (GeneID 151636). | |
0.1 mL | |
Zinc Finger | |
151636 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction