Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DTX3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552708
Description
Immunogen sequence: ERKPLSQQER DSCISPSEPE TKAEQKSNYF EVPLPYFEYF KYICPDKINS IEKRFGVNIE IQESSPNMVC LDFTSSRSGD LEAARESFAS EFQKNTEPLK QECVSLADSK QANKFKQELN HQFTKL Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 46%.
Component of the shelterin complex (telosome) that is involved in the regulation of telomere length and protection. Shelterin associates with arrays of double-stranded TTAGGG repeats added by telomerase and protects chromosome ends. Without its protective activity, telomeres are no longer hidden from the DNA damage surveillance and chromosome ends are inappropriately processed by DNA repair pathways. Promotes binding of POT1 to single-stranded telomeric DNA. Modulates the inhibitory effects of POT1 on telomere elongation. The ACD-POT1 heterodimer enhances telomere elongation by recruiting telomerase to telomeres and increasing its processivity. May play a role in organogenesis.
Specifications
| DTX3L | |
| Polyclonal | |
| Unconjugated | |
| DTX3L | |
| AU042200; Bbap; BC023741; B-lymphoma- and BAL-associated protein; deltex 3 like, E3 ubiquitin ligase; deltex 3-like; deltex 3-like (Drosophila); deltex E3 ubiquitin ligase 3L; DTX3L; E3 ubiquitin-protein ligase DTX3L; Protein deltex-3-like; RGD1565144; rhysin 2; Rhysin2; rhysin-2; RING-type E3 ubiquitin transferase DTX3L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 151636 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TDB6 | |
| DTX3L | |
| Recombinant protein corresponding to Human DTX3L. Recombinant protein control fragment (Product #RP-89860). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction