Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTX4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549652

Catalog No. PA549652


Only null left
Add to Cart

Description

Description

Predicted to react with mouse based on sequence homology.

DTX4 is a regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. It functions as an ubiquitin ligase protein in vivo, mediating 'Lys48'-linked polyubiquitination and promoting degradation of TBK1, targeting to TBK1 requires interaction with NLRP4.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DTX4
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q9Y2E6
DTX4
KLH conjugated synthetic peptide between 56-83 amino acids from the N-terminal region of human DTX4.
400 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DTX4
AI449438; BC044798; D930042H13; deltex 4 homolog; deltex 4 homolog (Drosophila); deltex 4, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 4; deltex homolog 4; deltex4; DTX4; E3 ubiquitin-protein ligase DTX4; KIAA0937; LRRGT00037; Protein deltex-4; RING finger protein 155; RING-type E3 ubiquitin transferase DTX4; RNF155
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
23220
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.