Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DTX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563346 Supplier Invitrogen™ Supplier No. PA563346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSPPMSPCSC PQCVLVMSVK AAVVNGSTGP LQLPVTRKNM PPPGVVKLP Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.

DTX4 is a regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. It functions as an ubiquitin ligase protein in vivo, mediating 'Lys48'-linked polyubiquitination and promoting degradation of TBK1, targeting to TBK1 requires interaction with NLRP4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DTX4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DTX4
Gene Accession No. Q9Y2E6
Gene Alias AI449438; BC044798; D930042H13; deltex 4 homolog; deltex 4 homolog (Drosophila); deltex 4, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 4; deltex homolog 4; deltex4; DTX4; E3 ubiquitin-protein ligase DTX4; KIAA0937; LRRGT00037; Protein deltex-4; RING finger protein 155; RING-type E3 ubiquitin transferase DTX4; RNF155
Gene Symbols DTX4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DTX4. Recombinant protein control fragment (Product #RP-101733).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23220
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.