Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DTX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572842
Description
DTX4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
DTX4 is a regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. It functions as an ubiquitin ligase protein in vivo, mediating 'Lys48'-linked polyubiquitination and promoting degradation of TBK1, targeting to TBK1 requires interaction with NLRP4.
Specifications
DTX4 | |
Polyclonal | |
Unconjugated | |
DTX4 | |
AI449438; BC044798; D930042H13; deltex 4 homolog; deltex 4 homolog (Drosophila); deltex 4, E3 ubiquitin ligase; deltex E3 ubiquitin ligase 4; deltex homolog 4; deltex4; DTX4; E3 ubiquitin-protein ligase DTX4; KIAA0937; LRRGT00037; Protein deltex-4; RING finger protein 155; RING-type E3 ubiquitin transferase DTX4; RNF155 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
207521, 23220 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q6PDK8, Q9Y2E6 | |
DTX4 | |
DTX4 antibody for a 19 amino acid peptide near the center of human DTX4. The immunogen is within amino acids 230 - 280 of DTX4. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction