Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DTYMK Antibody (SR1364), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32250050UL
Description
DTYMK Monoclonal antibody specifically detects DTYMK in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
DTYMK | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1364 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
CDC8TMPKTYMKPP3731, deoxythymidylate kinase (thymidylate kinase), dTMP kinase, EC 2.7.4.9, FLJ44192, MGC198617, thymidylate kinase | |
A synthesized peptide derived from human DTYMK (Uniprot #: P23919) | |
50 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
1841 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction