Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DTYMK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560092
Description
Immunogen sequence: RGALIVLEGV DRAGKSTQSR KLVEALCAAG HRAELLRFPE RSTEIGKLLS SYLQKKSDVE DHSVHLLFSA NRWEQVPLIK EKLSQGVTLV VDRYAFSGVA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
Thymidylate kinase (DTYMK) catalyzes the phosphorylation of dTMP to form dTDP in the dTTP synthesis pathway for DNA synthesis. DTYMK is essential for DNA synthesis and is an important intermediate enzyme in the pathway of many pyrimidine analog drugs, including 5-Fluorouracil (5-FU), the chemotherapeutic drug of choice for several solid tumors. The human DTYMK gene was isolated via functional complementation of a Saccharomyces cerevisiae cell cycle mutant, cdc8. DTYMK mRNA levels and enzyme activities are regulated at the level of cell cycle and cell growth pathways.
Specifications
DTYMK | |
Polyclonal | |
Unconjugated | |
DTYMK | |
AU044245; CDC8; D10Ertd203e; deoxythymidylate kinase; deoxythymidylate kinase (thymidylate kinase); dTMP kinase; DTYMK; mtmk; PP3731; Thymidylate kinase; Tmk; TMPK; TYMK; TYPK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P23919 | |
DTYMK | |
Recombinant protein corresponding to Human DTYMK. Recombinant protein control fragment (Product #RP-98924). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction