Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heater, Dual block
SCI_ED

Catalog No. s28682 Shop All Fisherbrand Products
Click to view available options
Temperature Range (Metric):
Ambient +5° to 130°C
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Dry bath, US-style plug, block lifter. (Blocks are sold separately.)

Offers a range of configurations with interchangeable modular blocks for a variety of application needs

  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment
  • Note: Blocks are sold separately

Applications:

Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation

Warranty and Services

2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, cULus, RoHS
Controller Type PID
Plug Type US
Relative Humidity <80%
Temperature Range (Metric) Ambient +5° to 130°C
Type Dry Bath/Block Heater
Fuse 250V 5A
Heat Up Time ≤ 20 min. 30° to 130°C
Includes Dry bath, US style plug, block lifter
No. of Blocks 2
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 1°C
Timer 0 to 99:59 min or continuous
Dimensions (L x W x H) 12.5 x 7.9 x 3.9 in. (318 x 200 x 100 mm)
Weight (English) 7.28 lb.
Weight (Metric) 3.3 kg
Electrical Requirements 100/120 V, 50/60 Hz
Voltage 100/120 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.