Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allegro™ Dual-Flex Wrist Support

Catalog No. p-93689
Click to view available options
Size:
Small
Large
X-Large

Catalog No. 19-037-866D

Supplier: Allegro™ 721203

Only null left
Add to Cart
Add to Cart

Specifications

Color Black
Closure Type Hook and loop
Product Type Dual-Flex™ Wrist Support
Size Large
For Use With (Application) Offers firm support limit flexion (downward) and extension (upward) movement and encourages natural wrist position
Includes Four stays on the front and back
Hand Ambidextrous
Material Nylon
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.