Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allegro™ Dual-Flex Wrist Support
Click to view available options
Size:
Small
Large
X-Large
Specifications
Specifications
Color | Black |
Closure Type | Hook and loop |
Product Type | Dual-Flex™ Wrist Support |
Size | X-Large |
For Use With (Application) | Offers firm support limit flexion (downward) and extension (upward) movement and encourages natural wrist position |
Includes | Four stays on the front and back |
Hand | Ambidextrous |
Material | Nylon |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction