Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ducks Unlimited™ Hearing and Safety Eyewear Combo Kit

Combo Kit includes one pair of PM8010 Ear Muffs along with one pair of Venture™ 3 Safety Glasses.

Specifications

Color Black, Gray
ANSI S3.1974 Yes
Cushion Material Foam
Cushion Color Black
For Use With (Application) Variety of industrial and recreational applications.
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-155-302
View Documents
Pyramex™
DUCOMBO5710
Ear Muffs and Clear Shooting Glasses N/A N/A N/A
19-155-303
View Documents
Pyramex™
DUCOMBO5740
Ear Muffs and Orange Shooting Glasses N/A N/A N/A
Description

Description

  • Convenient Combo Kit includes one pair of ear muffs and one pair of safety glasses
  • Venture™ Eyewear features soft, adjustable, ventilated nose piece
  • Eyewear frame is ventilated to prevent fogging
  • Lens is scratch resistant
  • Ear Muff features low-profile design with soft ear cups
  • Fold-away padded headband for comfortability and easy storage
Specifications

Specifications

Black, Gray
Foam
Variety of industrial and recreational applications.
26 dB
Black
Yes
Black
1 Ea. (Ear Muff/Safety Eyewear)
Combo Kit
Over-the-head
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.