Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DULLARD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557951
Description
Immunogen sequence: YIKDLSVVHS DLSSIVILDN SPGAYRSHPD NAIPIKSWFS DPSDTALLNL LPMLDALRFT ADVRSVLS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Serine/threonine protein phosphatase forming with CNEP1R1 an active phosphatase complex that dephosphorylates and may activate LPIN1 and LPIN2. LPIN1 and LPIN2 are phosphatidate phosphatases that catalyze the conversion of phosphatidic acid to diacylglycerol and control the metabolism of fatty acids at different levels. May indirectly modulate the lipid composition of nuclear and/or endoplasmic reticulum membranes and be required for proper nuclear membrane morphology and/or dynamics. May also indirectly regulate the production of lipid droplets and triacylglycerol. May antagonize BMP signaling.
Specifications
DULLARD | |
Polyclonal | |
Unconjugated | |
CTDNEP1 | |
2610507E10Rik; CTD nuclear envelope phosphatase 1; CTDNEP1; C-terminal domain nuclear envelope phosphatase 1; Dullard; dullard homolog; HSA011916; NET56; serine/threonine-protein phosphatase dullard | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23399 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95476 | |
CTDNEP1 | |
Recombinant protein corresponding to Human DULLARD. Recombinant protein control fragment (Product #RP-95710). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction