Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Duloxetine hydrochloride
333.87 g/mol
Specifications
Molecular Weight (g/mol) | 333.87 |
---|---|
Product Type | Duloxetine hydrochloride |
Solubility Information | Solubility aq. soln.: .. Other solubilities: soluble in methanol |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC463140010
|
Thermo Scientific Chemicals
463140010 |
1 g | N/A | N/A | N/A | ||||
AC463140050
|
Thermo Scientific Chemicals
463140050 |
5 g | N/A | N/A | N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
333.87 | |
Duloxetine hydrochloride | |
Solubility aq. soln.: .. Other solubilities: soluble in methanol |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title