Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Duolite™ C 206A, Thermo Scientific™

Catalog No. AC276630010
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle

 

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00146795

Specifications

Color Brown
Packaging Plastic Bottle
Quantity 1 kg
Physical Form Beads
Chemical Name or Material Duolite™ C 206A

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.