Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ DuoSet ELISA Ancillary Reagent Kit 1

Catalog No. p-200085174
Click to view available options
Quantity:
1 Kit (for 5 Plates)
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.