Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585452
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585452 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585452 Supplier Invitrogen™ Supplier No. PA585452
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (89%), Rat (87%), Dog (86%), Pig (90%), Bovine (90%). Positive Control: A549, H1299, HCT116, unboiled A431, A431 membrane extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a glycoprotein and a member of the NADPH oxidase family. The synthesis of thyroid hormone is catalyzed by a protein complex located at the apical membrane of thyroid follicular cells. This complex contains an iodide transporter, thyroperoxidase, and a peroxide generating system that includes this encoded protein and DUOX2. This protein is known as dual oxidase because it has both a peroxidase homology domain and a gp91phox domain. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUOX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene DUOX1
Gene Accession No. Q9NRD9
Gene Alias 9930101G15Rik; AW987690; Dual oxidase 1; dual oxidase 2; DUOX; DUOX1; Duox2; flavoprotein NADPH oxidase; large NOX 1; LNOX1; LNOX2; long NOX 1; NADPH thyroid oxidase 1; nicotinamide adenine dinucleotide phosphate oxidase; NOXEF1; NOXEF2; P138-TOX; THOX1; THOX2; thyroid oxidase 1
Gene Symbols DUOX1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human DUOX1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53905
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.