Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DUOX2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061576G
Description
DUOX2 Polyclonal specifically detects DUOX2 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
DUOX2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated | |
Q9NRD8 | |
DUOX2 | |
Synthetic peptide made to an internal portion of human DUOX2 (within residues 400-500). [Swiss-Prot# Q9NRD8] | |
0.1 mL | |
Cell Biology, Lipid and Metabolism, Neuroscience | |
50506 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
dual oxidase 2, dual oxidase-like domains 2, EC 1.11.1.-, EC 1.6.3.1, flavoprotein NADPH oxidase, Large NOX 2, LNOX2P138(TOX), Long NOX 2, NADH/NADPH thyroid oxidase p138-tox, NADPH oxidase/peroxidase DUOX2, NADPH thyroid oxidase 2, nicotinamide adenine dinucleotide phosphate oxidase, NOXEF2, p138 thyroid oxidase, P138-TOX, THOX2TDH6, Thyroid oxidase 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction