Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DUOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571497
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571497 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571497 Supplier Invitrogen™ Supplier No. PA571497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DUOX2 Polyclonal Antibody for Western Blot, Flow

The protein encoded by this gene is a glycoprotein and a member of the NADPH oxidase family. The synthesis of thyroid hormone is catalyzed by a protein complex located at the apical membrane of thyroid follicular cells. This complex contains an iodide transporter, thyroperoxidase, and a peroxide generating system that includes this encoded protein and DUOX1. This protein is known as dual oxidase because it has both a peroxidase homology domain and a gp91phox domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUOX2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene DUOX2
Gene Accession No. Q9NRD8
Gene Alias Dual oxidase 2; dual oxidase-like domains 2; DUOX2; flavoprotein NADPH oxidase; large NOX 2; LNOX2; Long NOX 2; NADH/NADPH thyroid oxidase p138-tox; NADPH oxidase/peroxidase DUOX2; NADPH thyroid oxidase 2; nicotinamide adenine dinucleotide phosphate oxidase; NOXEF2; p138 thyroid oxidase; P138-TOX; TDH6; THOX2; Thyroid oxidase 2
Gene Symbols DUOX2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 513-542 amino acids from the Central region of human DUOX2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50506
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.