Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DUOX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597677
Description
DUOX2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The protein encoded by this gene is a glycoprotein and a member of the NADPH oxidase family. The synthesis of thyroid hormone is catalyzed by a protein complex located at the apical membrane of thyroid follicular cells. This complex contains an iodide transporter, thyroperoxidase, and a peroxide generating system that includes this encoded protein and DUOX1. This protein is known as dual oxidase because it has both a peroxidase homology domain and a gp91phox domain.
Specifications
| DUOX2 | |
| Polyclonal | |
| Unconjugated | |
| DUOX2 | |
| Dual oxidase 2; dual oxidase-like domains 2; DUOX2; flavoprotein NADPH oxidase; large NOX 2; LNOX2; Long NOX 2; NADH/NADPH thyroid oxidase p138-tox; NADPH oxidase/peroxidase DUOX2; NADPH thyroid oxidase 2; nicotinamide adenine dinucleotide phosphate oxidase; NOXEF2; p138 thyroid oxidase; P138-TOX; TDH6; THOX2; Thyroid oxidase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50506 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.837 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4 | |
| Q9NRD8 | |
| DUOX2 | |
| Recombinant Human Dual oxidase 2 protein (1-100AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction