Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUPD1 Monoclonal Antibody (OTI6C6), TrueMAB™, OriGene
SDP

Catalog No. 502541945
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1945 30 μL
50-167-2988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1945 Supplier OriGene Supplier No. TA501880S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DUPD1 Monoclonal antibody specifically detects DUPD1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen DUPD1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SLFNL1
Gene Accession No. Q499Z3
Gene Alias atypical dual-specific protein phosphatase; dual specificity phosphatase 27; dual specificity phosphatase and pro isomerase domain containing 1; dual specificity phosphatase DUPD1; DUPD1; DUSP27; FMDSP
Host Species Mouse
Immunogen Full length human recombinant protein of human SLFNL1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200172
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.