Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUPD1 Mouse anti-Human, Clone: OTI7E4, lyophilized, TrueMAB™
SDP

Catalog No. 501661019
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-1019 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-1019 Supplier OriGene Supplier No. CF502080
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

DUSP27 (Dual Specificity Phosphatase 27, Atypical) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include phosphatase activity and protein tyrosine/serine/threonine phosphatase activity. An important paralog of this gene is DUSP13.

Specifications

Antigen DUPD1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7E4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DUPD1
Gene Accession No. Q68J44
Gene Alias DUSP27, FMDSP
Gene Symbols DUPD1
Host Species Mouse
Immunogen Full length human recombit protein of human DUPD1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338599
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.