Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ 270 Series Sterile Frocks
BIOPHARMAGREENER_CHOICE

Catalog No. 190881127D Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Length (English):
39 in.
40 in.
41 in.
43 in.
44 in.
45 in.
46 in.
Length (Metric):
99 cm
101.6 cm
104.14 cm
109.2 cm
111.76 cm
114.3 cm
116.8 cm
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Clean-processed and sterile frocks provide an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

  • Garments are gamma sterilized to SAL of 10-6
  • Body-length jacket design with front snap closures for easy donning and doffing
  • Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
  • Unique, patented flashspinning process creates an barrier to dry particles, microorganisms, and nonhazardous liquids
  • Garments are constructed with the smooth side out to reduce particulate entrapment and bound seams reduce lint
  • ISO Class 5 (formerly Class 100) compatible
  • Processed in a certified ISO Class 4 (Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
  • Generous sizing permits freedom of movement
  • Plastic snaps won't rust or flake during processing
  • Bound seams are covered with garment fabric to reduce the potential for particle penetration
  • Elastic opening for tighter fit at wrist
  • Bound neck for lower particle shedding, adjustable snap at neck for better fit
  • Full traceability
TRUSTED_SUSTAINABILITY

Specifications

Product Type Controlled Environments Frock
Size X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Sterile
Closure Type Snaps
Cuff Style Elastic
Seam Type Bound
No. of Pockets None
Style IsoClean™ 270 Series
Size (English) Chest 38 to 40 in.
Size (Metric) Chest 96 to 101 cm
Sleeve Style Long Sleeve
Packaging Type Clean-processed, Individually Packaged
Length (English) 43 in.
Length (Metric) 109.2 cm
Length (English) Sleeve 37.75 in.
Length (Metric) Sleeve 95.8 cm
Cleanroom Class Class 100 (ISO 5)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.