Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 60 Coveralls, Attached Hood and Boots

Catalog No. 19067056 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Height (English):
5 to 5.7 ft.
5.3 to 5.7 ft.
5.5 to 5.9 ft.
5.8 to 6.2 ft.
6 to 6.4 ft.
6.2 to 6.4 ft.
6.4 to 6.7 ft.
6.7 to 6.10 ft.
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls with attached skid-resistant boots are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.

  • Lightweight coveralls (1.8 oz. per sq. yd.) provide barrier protection against nontoxic particle, liquid, and spray hazards.
  • Microporous film that is laminated to a spundbonded polypropylene substrate.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements.
  • Attached hood with elastic around face opening.
  • Front zipper closure for easy donning and doffing.
  • Elastic opening for tighter fit at wrist
  • Attached ProShield 3 boots have special coating to provide added skid-resistance.
  • Manufactured under specifications that do not contain natural rubber latex.

Compliance

Meets USDA, FDA, OSHA standards; ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.

  1. Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
  2. ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
  3. Certified incinerable by independent laboratory. For details, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 6X-Large
Gender Unisex
Material Non-woven Polypropylene Substrate, Microporous Film Laminate
Color White
Disposable Single-use
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard, Elastic Face
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle, Liquid
Certifications/Compliance USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Description Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Latex Free Yes
Model NG120SWH
For Use With (Application) Automotive Refinishing, Waste Cleanup, Asbestos Abatement, Hazardous Remediation
Quantity 25/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.