Learn More
DuPont™ ProClean™ Limited-Use Cleanroom Frocks
Catalog No. 190881018 Shop All DuPont Personal Protection Products
Click to view available options
Closure Type:
Snaps
Zipper
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Maximum containment short of a full coverage coverall. DuPont™ ProClean™ limited-use, full-length frocks offer the breathability and comfort of microporous film fabric.
Proprietary polypropylene laminate fabric provides effective contamination control and protection from liquid-splash and dry-particulate cleanroom hazards.
  • White knee-length frocks
  • Microporous film fabric
  • Serged seams
  • Elastic wrist
  • Bound neck
  • Available with a five-snap or zipper front closure
  • Antistatic treated
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Frock
Size X-Large
Material Microporous Composite Fabric
Color White
Sterility Non-sterile
Closure Type Zipper
Cuff Style Elastic
Seam Type Serged
Cleanroom Class Class 1000 (ISO 6)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.