Learn More
DuPont™ ProClean™ Frock
Catalog No. 190881033 Shop All DuPont Personal Protection Products
Click to view available options
Closure Type:
Snaps
Zipper
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
Catalog No. 19-088-1033
Supplier: DuPont™ PC270SWHSM00300B
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Maximum containment short of a full coverage coverall. DuPont™ ProClean™ limited-use, full-length frocks offer the breathability and comfort of microporous film fabric.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Elastic opening for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Bulk packaged in double transparent poly liners.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Frock
Size Small
Material Microporous Composite Fabric
Color White
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Seam Type Serged
Cleanroom Class Class 1000 (ISO 6)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.