Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 10 Coveralls, Attached Hood and Skid-Resistant Boots

Catalog No. 19162517 Shop All DuPont Personal Protection Products
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.

  • Comfort fit design based on extensive wearer input provides a comfortable garment design that enables a greater range of movement while stretching and bending, a more tailored fit, reinforcement in high stress areas for fewer blowouts, a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Attached hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Attached skid-resistant boots
  • White or blue
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Sterility Non-sterile
Certifications/Compliance ASTM D1117
Model PB122S
For Use With (Application) General Maintenance, Janitorial, Cleaning
Quantity 25/Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.