Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ ProShield™ 10 Coveralls, Elastic Wrists and Ankles
Shop All DuPont Personal Protection Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Color:
Blue
White
Description
Spunbond meltblown spunbond (SMS) coveralls protect against non-hazardous particles and light liquid splashes in a variety of industries.
- Soft, breathable and lightweight with comfort fit design
- Spunbond Meltblown Spunbond (SMS)
- Disposable
- Laydown collar
- Elastic wrists, ankles
- Front zipper closure
- Serged seams
- Available in blue or white
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Collared |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction