Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 60 Boot Covers

Catalog No. 19715314 Shop All DuPont Personal Protection Products
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy

Catalog No. 19-715-314

Supplier: DuPont™ NG456SWHMD010000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Boot covers with PVC skid-resistant soles are suitable for protection against non-hazardous particles, light liquid splash, and aerosols in many industrial applications.

  • 18 in. high boot cover with elastic top provides coverage up to calf
  • PVC soles provides enhanced skid-resistance and durability
  • Manufactured under specifications that do not contain natural rubber latex
  • One size fits most
TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Material ProShield
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Top
Sole Type Non-skid
Model NG456SWHMD010000
Seam Type Serged
Thickness 10 mil
For Use With (Application) Automotive, Refinishing, Waste Cleanup, Remediation
Length (English) 11.75 in.
Length (Metric) 29.84 cm
Height (English) 18 in.
Height (Metric) 45.72 cm
Resistance Dry Particulates, Fluid
Product Type Boot Covers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.