Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ 30 Shoe Covers

Catalog No. 19047744 Shop All DuPont Personal Protection Products
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Choose from blue or white slip-resistant sole shoe covers suitable for Cleanroom ISO 6 (Class 1000) controlled environments.

  • Slip resistant sole shoe covers
  • Available in blue or white
  • Generously sized in large or x-large
  • Resistant to liquids
  • Will not delaminate
  • Lint-free
  • Elastic ankles for secure fit
  • Seamless
  • Sold in case of 100 pairs
  • Federal 209E Cleanroom Class 1,000
  • Protects against non-hazardous dry particulates, liquids, and powders
  • ISO Cleanroom Class ISO 6 (Class 1,000) and above
  • Material polyethylene coated spunbond polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Size Large
Material Spunbond Polypropylene
Latex Free Yes
Coating Polyethylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Anti-skid
Seam Type Serged
Certifications/Compliance Federal 209E, ASTM D3776
Packaging Quantity 100 Pairs/Cs.
Packaging Type Bulk
Length (English) 14.75 in.
Length (Metric) 37.46 cm
Height (English) 5.5 in.
Height (Metric) 13.97 cm
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 1000 (ISO 6)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.