Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 9000 Coveralls

Catalog No. 19715249
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Ankle Style:
Attached Socks, Boot Overflaps
Elastic
This item is not returnable. View return policy
This item is not returnable. View return policy

For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams and utilities

Tychem BR fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents.

  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Strong zipper for tough, flexible closure
  • Elastic opening for tighter fit at wrist
  • Yellow

Model BR127TYL

  • Three-piece hood design with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Elastic opening for tighter fit at ankle
Model BR128TYL
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Longer zipper extends to chin for complete coverage of neck area
  • Integrated socks composed of garment material

Compliance

Meets ASTM™ D1777N, ASTM D3776, ASTM D3786, ASTM D5034; 16 CFR 1610 Standard for the Flammability of Clothing Textiles.

TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Material Tychem
Color Yellow
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Taped
Description Respirator fit hood, elastic wrist, attached socks with outer boot flaps; Multilayer film barrier laminated to Polypropylene substrate
Size (English) Chest 36.25 to 39.75 in.
Size (English) Inseam 28.5 in.
Size (Metric) Chest 92.07 to 100.96 cm
Size (Metric) Inseam 72.39 cm
Thickness 16 mil
For Use With (Application) Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
Length (English) Sleeve 34.75 in.
Length (Metric) Sleeve 88.26 cm
Width (English) Chest 24.75 in.
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.