Learn More
DuPont™ Tychem™ BR Encapsulated Suit
Shop All DuPont Personal Protection ProductsDescription
Tychem BR fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Integrated socks composed of garment material
- Standard 20 mil PVC faceshield
- Elastic opening for tighter fit at wrist
- Yellow
Model BR527TYL
Front entry design allows wearer to partially don garment (maintain standby position) without assistance
Model BR528TYL
Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams, and utilities

Specifications
Specifications
Color | Yellow |
For Use With (Application) | Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities |
Material | Tychem |
Size | Large |
Closure Type | Hook and Loop |
Description | Encapsulated level B suit, expanded back, elastic wrist, attached socks with outer boot flaps, double storm flap; Multilayer film barrier laminated to Polypropylene substrate |
Height (English) | 5.9 to 6.3 ft. |
Height (Metric) | 1.79 to 1.92 m |
Length (English) Inseam | 30.75 in. |
Length (English) Sleeve | 29.25 in. |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.