Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ BR Encapsulated Suit

Catalog No. p-4931083 Shop All DuPont Personal Protection Products
Click to view available options
Closure Type:
Hook and Loop
Description:
Encapsulated level B suit, expanded back, elastic wrist, attached socks with outer boot flaps, double storm flap; Multilayer film barrier laminated to Polypropylene substrate
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment

Tychem BR fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents

  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Integrated socks composed of garment material
  • Standard 20 mil PVC faceshield
  • Elastic opening for tighter fit at wrist
  • Yellow

Model BR527TYL
Front entry design allows wearer to partially don garment (maintain standby position) without assistance

Model BR528TYL
Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment

For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams, and utilities

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
For Use With (Application) Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities
Material Tychem
Size X-Large
Closure Type Hook and Loop
Description Encapsulated level B suit, expanded back, elastic wrist, attached socks with outer boot flaps, double storm flap; Multilayer film barrier laminated to Polypropylene substrate
Height (English) 5.9 to 6.3 ft.
Height (Metric) 1.79 to 1.92 m
Length (English) Inseam 30.75 in.
Length (English) Sleeve 29.25 in.
Length (Metric) Sleeve 74.2 cm
Seam Type Taped
Size (English) Chest 30 in.
Size (Metric) Chest 118.74 to 127.63 cm
Thickness 16 mil
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.