Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ CPF ™ 2 Coveralls

Catalog No. 19088208 Shop All DuPont Personal Protection Products
Click to view available options
Color:
Tan
Description:
Standard fit hood, elastic wrist and ankle; Multilayer composite barrier film laminated to a durable Polypropylene substrate
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Great tear resistance

  • Provide greater chemical holdout, offering protection from a wide variety of chemicals
  • Rugged and durable even in environments where abrasion is an issue
  • Fabric consists of Saranex™ 23-P barrier film on tough 1.5 oz. per sq. yd. polypropylene substrate
  • Gray

TRUSTED_SUSTAINABILITY

Specifications

Size X-Large
Material Tychem
Color Tan
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper
Seam Type Taped
Description Standard fit hood, elastic wrist and ankle; Multilayer composite barrier film laminated to a durable Polypropylene substrate
Size (English) Inseam 29.5 in.
Size (Metric) Chest 104.77 to 113.66 cm
For Use With (Application) Chemical handling, petrochemical, hazardous materials, waste cleanup, fire departments, industrial hazmat teams, utilities and domestic preparedness
Length (English) Sleeve 35.5 in.
Length (Metric) Sleeve 90.17 cm
Width (English) Chest 27.25 in.
Height (English) 5.8 to 6.2 ft.
Height (Metric) 1.76 to 1.88 m
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.