Learn More
DuPont™ Tychem ™ 5000 Coveralls with Hood and Overboots/Socks
Shop All DuPont Personal Protection ProductsDescription
Provide defense for applications such as acid handling, tank cleaning, chemical handling, petrochemical market, hazardous waste cleanup, industrial hazmat teams and fire department and utilities use.
- Zipper front with attached hood and elastic wrists and overboots
- Multilayer barrier film laminated to a lasting 2 oz. per sq. yd.
- Polypropylene substrate offers greater physical strength and chemical holdout
- Set sleeve pattern for greater arm movement
- Coveralls have front-entry zipper
- Tan
Compliance
Meets ANSI™/ISEA™ 101-1996 standards; complies with CAN/CGSB-155.20-2000 for flame resistance; NFPA 701 Standard Methods for Fire Tests for Flame Propagation of Textiles and Films (1989); NFPA 99 Health Care Facilities requirements.
†Attached socks are to be worn inside the user's own chemical protective boots. The boot splash cover adds additional protection. These attached socks do not have adequate durability or slip resistance to be worn as outer footwear covering.

Specifications
Specifications
Size | X-Large |
Material | Tychem |
Color | Tan |
Disposable | Single-use |
Ankle Style | Attached Socks |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Attached, Elastic Face |
Closure Type | Zipper with Storm Flap |
Seam Type | Taped |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.