Learn More
DuPont™ Tychem™ 6000 Taped Seam Coveralls
Shop All DuPont Personal Protection Products
Click to view available options
Size:
Small
3X-Large
5X-Large
6X-Large
7X-Large
Description
- Strong and durable coverall with broad chemical barrier
- Tychem™ 6000 coverall is made of barrier film laminated to Tyvek™ substrate
- For use when potential exposure to industrial chemicals and chemical warfare agents exists
- Provides protection against over 180 chemical challenges for at least 30 minutes
- Available in low-visibility gray fabric for discretionary purposes
- With taped seams and attached hood (respirator fit)
- Front zipper closure and storm flap with tape closure
- Elastic wrists and elastic ankles
Specifications
Specifications
| Size | 6X-Large |
| Material | Tychem |
| Color | Gray |
| Seam Type | Taped |