Learn More
DuPont™ Tychem™ QC 120 Series Coveralls
Catalog No. 19041747
Click to view available options
Seam Type:
Bound
Serged
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Flexible and durable light liquid splash protection
Tychem™ QC is polyethylene-coated Tyvek™ fabric fabric provides at least 30 minutes of protection against 42 chemical challenges
  • Laydown collar
  • Open wrists
  • Open ankles
  • Yellow for high visibility
  • 10 mil thickness
QC 120B
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
QC 120S
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seams
  • QC 120T
    • Taped seams provide strong chemical resistance against heavy liquid splashes
    • A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
    TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Collared Coveralls
    Size Large
    Material Tychem
    Color Yellow
    Ankle Style Open
    Cuff Style Open
    Seam Type Bound
    Collar Type Laydown
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.