Learn More
DuPont™ Tychem™ 10,000FR Chemical Suit
Description
Totally encapsulating Level A suit is a multi-layer fabric that passes the ASTM™ F 1001 test battery standards.
- Seams are sewn and triple heat-sealed inside
- Features front entry, expanded back, exhaust valves, gastight zipper and attached socks† with boot flap
- Gloves made of Barrier™, neoprene and Kevlar™ knit
- 40mil PVC faceshield has 5mil PTFE overlay
Compliance
Meets NFPA 1991 Standard on Vapor Protective Ensembles for Hazardous Materials Emergencies and CBRN Terrorism Incident.
Specifications
Specifications
| Certifications/Compliance | NFPA 1991 |
| Color | Silver |
| For Use With (Application) | Ideal for industrial chemical, HazMat response and military applications |
| Material | Tychem |
| Size | X-Large |
| Ankle Style | Attached socks, Boot overflaps |
| Closure Type | Front zipper |
| Cuff Style | Attached gloves |
| Description | Standard width faceshield, 2 exhaust valves, internal adjustment waist belt, attached internal North Silver Shield™/4H™/external Neoprene size 10 gloves, separate Dupont™ Kevlar™ aramid fiber knit over gloves, socks with outer boot flaps, soles |
| Height (English) | 5.9 to 6.3 ft. |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
